Elatusainepullo, neliönmuotoiset, kierrekorkillinen, Nalgene®
Toimittaja: Thermo Fisher Scientific
322020-0125 322020-0030 322020-0060 322020-0250 322020-0500 322020-1000 322020-2000
NALG322020-0125EA 227 EUR
NALG322020-0125 NALG322020-0030 216-2987 NALG322020-0250 NALG322020-0500 NALG322020-1000 NALG322020-2000
Elatusainepullo, neliönmuotoiset, kierrekorkillinen, Nalgene®
Pullot Väliainepullot
PETG bottle with HDPE screw closure.
  • Graduated
  • Durable
  • Break-resistant
  • Leakproof
Package and ship liquid media, buffers and sera using square media bottles with screw caps. Heavy-walled, durable bottles offer excellent gas barrier properties and are specifically designed for storing media and sera. Non-sterile bottle offers excellent gas barrier properties; clear PETG material allows for easy identification of contents.
Sertifikaatit: USP Class VI, FDA Compliance 21 CFR