Pullo, kapea suuaukko, diagnostiset pullot, pyöreät, kierrekorkilla, Nalgene®
Toimittaja: Thermo Fisher Scientific
322006-9025 322006-9050 322006-9125
NALG322006-9025EA 2300 EUR
NALG322006-9025 NALG322006-9050 NALG322006-9125
Pullo, kapea suuaukko, diagnostiset pullot, pyöreät, kierrekorkilla, Nalgene®
Pullot Muovipullot
Bottles with polypropylene closure used for safely storage and ship small volume samples and valuable reagents.
  • Nonsterile
  • Made up of natural translucent PPCO
  • Leakproof
Thermo Scientific™ Nalgene™ Natural PPCO diagnostic bottles with closures are used in test kits and many small-volume applications. These autoclavable and durable polypropylene copolymer bottles are tray packed with closure for easy handling and feature uniform walls resistant to splitting or puncturing.
Sertifikaatit: Compliance with ISO 13485:2003 and 21CFR177.1520, USP class VI.