Koripullot, hygieeniset laipat, Nalgene®
Toimittaja: Thermo Fisher Scientific
2640-0020 2640-0050 2640-0130
NALG2640-0020EA 841 EUR
NALG2640-0020 NALG2640-0050 NALG2640-0130
Koripullot, hygieeniset laipat, Nalgene®
Pullot Jerrykannut
Made of PP, translucent.
  • Graduated to contain in liters and gallons
  • With thermoplastic elastomer gasket
  • Molded-in handles
  • Excellent chemical and temperature resistance
Use the autoclavable PP Carboys with 1,5" sanitary flanges as dispensing ports or with secure hookup for supply reservoirs to systems such as fermentors or chromatography columns.