Drawer cabinet, VISION 12
Toimittaja: MULTIROIR
VISION12
MULCVISION12EA 160 EUR
MULCVISION12
Drawer cabinet, VISION 12
Laatikot Säilytyslaatikot
Drawer cabinet VISION 12 contains 16 drawers made of clear plastic.
  • Dark grey frame
  • Transparent drawers
  • Can be wall mounted
W×D×H (single drawer): 43×102×44 mm
Weight: 1,10 kg