Stackable Storage Boxes, Polyethylene
Toimittaja: MULTIROIR
9276000000 9336000000 8531000
MULC9276000000EA 85.6 EUR
MULC9276000000 MULC9336000000 MULC8531000
Stackable Storage Boxes, Polyethylene
Laatikot Säilytyslaatikot
White polyethylene bins designed for laboratory environments. Smooth walls and rounded edges for easy, hygienic cleaning. Sturdy, stackable and available in three sizes. Suitable for intensive professional use.
  • Three sizes available
  • Closed handles with rounded edges
  • Smooth walls and rounded inner corners for optimal cleaning
  • Stackable to save space
  • Compatible with the requirements of laboratories and sensitive industries
  • Variable operating temperature depending on model
  • Solid walls/base
Made from high-quality polyethylene, these solid-sided white