Plastic Boxes, Safety Closure
Toimittaja: MULTIROIR
47509 47532 47533 47534 47535 47510
MULC47509EA 206 EUR
MULC47509 MULC47532 MULC47533 MULC47534 MULC47535 MULC47510
Plastic Boxes, Safety Closure
Laatikot Säilytyslaatikot
The plastic boxes with secure lid allow the storage of your liquid and solid products.
  • Safety closure
  • 100% liquid-tight
  • Highly impact-resistant PP material
  • Dishwasher safe. Microwave safe
  • Food contact
  • Material compatible with very low temperatures
  • Standard colour: Transparent
They offer a perfect seal and a temper-evident closure, especially for liquid products.