Stackable plastic trays
Toimittaja: MULTIROIR
26500PENPR 26501PENPR 26502PENPR 26503PENPR 26505PENPR 26506PENPR
MULC26500PENPREA 100 EUR
MULC26500PENPR MULC26501PENPR MULC26502PENPR MULC26503PENPR MULC26505PENPR MULC26506PENPR
Stackable plastic trays
Haihdutusmaljat/Tarjottimet/Altaat Laboratorioalustat/-astiat
Range of six stackable plastic trays for storage.
  • Made from polypropylene
  • Stackable
  • High impact resistance
  • Six colors available
Trays are suitable for use in temperatures ranging from −20 to 60 °C.