Plastic Box with White Lid
Toimittaja: MULTIROIR
25685
MULC25685EA 243 EUR
MULC25685
Plastic Box with White Lid
Laatikot Säilytyslaatikot
Box opaque with airtight and tamper-evident lid, perfect for use in laboratories.
  • Polypropylene material
  • Opaque white
  • Watertight lid
  • Tamper-proof lid
  • Stackable box
  • Food contact quality
Ideal for laboratory environments, this versatile container ensures the secure storage and transport of samples and reagents. The airtight, tamper-evident lid provides enhanced safety, while the stackable design maximises space efficiency. The opaque white polypropylene construction ensures durability and easy identification of contents.