Drawer storage, crystal multiroir M5000
Toimittaja: MULTIROIR
11108 11109 11158 11159 11128 11129
MULC11108EA 837 EUR
MULC11108 MULC11109 MULC11158 MULC11159 MULC11128 MULC11129
Drawer storage, crystal multiroir M5000
Laatikot Säilytyslaatikot
Crystal Multiroir M5000 storage boxes are modular, combinable and provided with a capacity of five litres.
  • Perfect transparency with possibility of autoclave sterilisation
  • High impact resistance with resistant to shocks
  • Available as a large drawer or 2 small drawers with the possibility to interchange all drawers