Pallets and sumps for IBC station
Toimittaja: asecos
8806 8807 8851 8854 19232
MSGA8806EA 1610 EUR
MSGA8806 MSGA8807 MSGA8851 MSGA8854 MSGA19232
Pallets and sumps for IBC station
Tynnyrivarasto
Steel, transportable.
  • Easy and safe in-house transport
  • Accessible from below
  • Robust construction
  • Available in painted and hot dip galvanised
For the safe storage of water pollutants and flammable liquids.