Combi safety storage cabinets, Type 90, K-UB-90
Toimittaja: asecos
30365001 30377001 30370001 30358001
135-1944EA 2710 EUR
135-1944 MSGA30377001 MSGA30370001 135-1777
Combi safety storage cabinets, Type 90, K-UB-90
Kalusteet Säilytyskaapit
Fireproof safety cabinets, classified type 90 (fire resistant up to 90 minutes), safe and approved storage of aggressive and flammable hazardous substances in working areas.
  • Robust construction and longevity - body made of powder-coated sheet steel
  • Two separated storage compartments for the storage of flammable liquids, acids and alkalis
  • User-friendly and safe - door and drawer remain open in any position, interior of the cabinet completely visible – access to all containers, 135° opening of the wing door
  • Safe - drawer fitted with earthing wire incl. clamp