Safety storage cabinets, type 90, S-PHOENIX Vol 2 - 90
Toimittaja: asecos
30315001 30315002 30316001 30316002 30119001 30235001 30235002 30236001 30236002 30119002
MSGA30315001EA 3280 EUR
MSGA30315001 MSGA30315002 MSGA30316001 MSGA30316002 135-1864 MSGA30235001 MSGA30235002 MSGA30236001 MSGA30236002 MSGA30119002
Safety storage cabinets, type 90, S-PHOENIX Vol 2 - 90
Kalusteet Säilytyskaapit
Fireproof safety cabinets, classified type 90 (fire resistant up to 90 minutes), safe, durable, robust and convenient.
  • Space saving folding doors
  • Safety elements assembled outside the storage compartment for increased protection against corrosion
  • One-hand operation for double-door cabinets
  • Door open arrest system, convenient and safe removal or storage of containers
  • Doors lockable with cylinder locking (integration in a locking system possible)
  • Adjusting aids to compensate for uneven floor
  • Integrated air ducts ready for connection (NW 75) to a technical