Safety dispensing cans
Toimittaja: asecos
16768 16769 16770
MSGA16768EA 255 EUR
MSGA16768 MSGA16769 MSGA16770
Safety dispensing cans
Turvakannut
Safety cans, made of high quality and chemically resistant stainless steel with PTFE gaskets. For safe and comfortable handling of small amounts of flammable liquids.
  • Easy handling, ergonomic design and high stability
  • Flame traps and explosion-proof
  • Pressure relief valve (vent at about 0,3 bar)
  • Self-closing metering devices
  • Moisteners for safe cleaning with solvents