Oil sorbents rolls
Toimittaja: asecos
11315 9936 9937
MSGA11315EA 283 EUR
MSGA11315 MSGA9936 MSGA9937
Oil sorbents rolls
Imeytysaineet ja imeytysliinat
Water repellent absorbent rolls for safe absorption of oil or petrol.
  • Capacity up to 16 times of its own weight
  • High rib and rub resistance
  • Perforated so that it needs only take sufficient material as required