Stereomikroskoopit, ST-30 ja ST-36
Toimittaja: MOTIC
51020141 PS30425201 PS36425201 PS3045B201 PS3645B201
632-9018EA 48.4 EUR
632-9018 630-3015 630-0691 MOTIPS3045B201 MOTIPS3645B201
Stereomikroskoopit, ST-30 ja ST-36
Mikroskoopit
These stereo microscopes are designed for education as well as quality control.
  • Bright, sharp image without heat
  • Magnification is changed by turning the turret
  • Comfortable long-term use, low consumption, environmental choice
  • Rear-facing head, forward-facing head or 360° rotatable head
  • Efficient LED energy saving lamp
Toimitustiedot: Supplied with frosted glass stage plates and dust cover.