Polarisaatiomikroskoopit, BA310-POL
Toimittaja: MOTIC
1100100402242 1101001703961 1101001703941 1101001703951 1101001703981 1101001703971
630-1822EA 4240 EUR
630-1822 MOTI1101001703961 MOTI1101001703941 MOTI1101001703951 MOTI1101001703981 MOTI1101001703971
Polarisaatiomikroskoopit, BA310-POL
Mikroskoopit
These models offer higher optical performance and modularity.
  • Polariser mounted under the condenser holder rotates 360° with click stop at 90°
  • Highly stable, robust symmetrical pyramid design and T-shaped base
  • Anti-fungal chemical treatment to inhibit germs growth
  • Plan achromatic, stain-free objectives
  • One eyepiece features a crosshair reticle
Suitable for life science, geology, material science and forensics.
Toimitustiedot: Supplied with an Allen key, a blue filter and a dust cover.