Metallurgical microscopes, PSM-1000
Toimittaja: MOTIC
1100101700071 1100101700062 SW010282A AP99777001 PP99777001
MOTI1100101700071EA 7020 EUR
MOTI1100101700071 MOTI1100101700062 MOTISW010282A MOTIAP99777001 MOTIPP99777001
Metallurgical microscopes, PSM-1000
Mikroskoopit
The PSM-1000 is a modular complex microscope.
  • Three-lens change-over turret to convert to laser work
  • Wavelength from 355 to 1064 nm achieved by optical lenses
  • Built-in camera port for documentation use, incident illumination available
  • Tilting head models for more ergonomic needs
  • Adjustable centrable, forward facing, quadruple nosepiece (dovetail mount) with one fixed and three adjustable positions
Designed for applications in the semiconductor industry, as well as for laser inspections and repairs, specially on wafers and printed circuit boards (PCB).