Vitrex Safety Pen Needles with Dual Automatic Protective Shields
Toimittaja: Vitrex Medical A/S
531113 531213 531313 531413
MODU531113EA 98.8 EUR
MODU531113 MODU531213 MODU531313 MODU531413
Vitrex Safety Pen Needles with Dual Automatic Protective Shields
Ruiskuneulat
Vitrex safety pen needles are intended for a safe and simple insulin injection via most insulin pens. The application is the same as normal insulin pens, but they offer an automatic double protection without extra operations after using. The coloured indicator shields in the front and back-end provide a visual, physical and audible confirmation of safety. The needles are sterile and individually packed.
  • Coloured automatic safety shields on front and back-end providing double protection after using
  • Triple sharpened needle and top lubrication technology,