Vitrex Ultrasafe Safety Blood Lancets
Toimittaja: Vitrex Medical A/S
393413 395413 396413 394413 398413 399413 392413
MODU393413EA 29.5 EUR
MODU393413 MODU395413 MODU396413 MODU394413 MODU398413 MODU399413 MODU392413
Vitrex Ultrasafe Safety Blood Lancets
Verilansetit
Sterile safety lancets intended for blood glucose and other in vitro diagnostic tests. Protects the health workers facing accidental needle-stick injury while caring for patients, putting themselves at risk to bloodborne illnesses including the AIDS virus and Hepatitis.
  • Pressure activated
  • Unique design, ergonomically optimised for better handling
  • Easy targeting of puncture site
High quality silicone coated needle: Less pain and smooth penetration.