Surface ATP test swabs, Clean-Trace™
Toimittaja: 3M Food Safety
UXL100
MMMPUXL100EA 417 EUR
MMMPUXL100
Surface ATP test swabs, Clean-Trace™
Vanupuikot Näytteenottopuikot näytteiden ottamiseen
Real-time adenosine tri-phosphate (ATP) surface hygiene test which gives an objective indication of cleanliness in seconds.
  • Self contained surface test designed to ensure samples can be collected in hard-to-reach places which are often difficult to clean
  • Metal detectable and blue plastic handle for safe use in a food manufacturing environment
  • A 30-second self-contained sanitation test to assess the standard of food production surfaces
With its unique liquid stable enzyme, the test offers superior levels of repeatability. This aspect of its performanceMore Product Information