Casting tape, Scotchcast™ Plus
Toimittaja: 3M MEDICAL
M82002A
MMMEM82002AEA 679 EUR
MMMEM82002A
Casting tape, Scotchcast™ Plus
Ensiaputarvikkeet Ensiapupakkauksen komponentit
This casting tape consists of a knitted fiberglass fabric impregnated with polyurethane resin. Exposure to moisture or water initiates a chemical reaction which causes the tape to become rigid.
  • Finished casts produced from this casting tape are lightweight, strong, radiolucent, and porous
  • Scotchcast™ Plus casting tape contains a lubricating agent in the resin to provide a slippery feel when exposed to water