Spore strips, Attest™
Toimittaja: 3M MEDICAL
M1262 M1264
MMMEM1262EA 733 EUR
MMMEM1262 MMMEM1264
Spore strips, Attest™
Sterilointi-indikaattori
3M™ Attest™ Biological Monitoring Systems are true biological indicators, in self-contained indicator capsules. They enable checks on the biological reliability and performance of each steriliser without laboratory facilities and laboratory trained personnel.
  • Self-contained, reducing the risk of false positives due to contamination
  • Colour change gives clear unambiguous, visual indication of PASS or FAIL
  • Time saving and results in 24 to 48 hours which require no interpretation