PELTOR™ X1 and X2 Earmuffs
Toimittaja: 3M
X1B-EU X2B-EU
MMMAX1B-EUEA 0 EUR
MMMAX1B-EU MMMAX2B-EU
PELTOR™ X1 and X2 Earmuffs
Kuulosuojaimet Hälyä estävät kuulosuojaimet
X series earmuffs are 3M’s latest advancement in hearing conservation. New technologies in comfort, design and protection all come together in this groundbreaking earmuff line. These lightweight, lower-profile earmuff designed for protection against low-to-moderate level industrial noise and other loud sounds.
  • Earmuff style: Behind the head
  • Material: ABS/Polyurethane
  • Design of neckband allows earmuffs to be worn with a variety of face shields and headgear
  • Compatible with headgear without accessory slots
  • Soft, adjustable head strap helps to maintain a