Face shield, WP98 series
Toimittaja: 3M
WP98
MMMAWP98EA 34.8 EUR
MMMAWP98
Face shield, WP98 series
Kasvosuojaimet
The WP98 Series Face Shield is a high impact resistant, clear acetate faceshield designed to fit 3M hard hats. It can be used with the H8 Headgear easy faceshield replacement system.
  • Provides protection against impacts, liquids, and chemical splashes
  • Clear acetate faceshield
  • High impact resistance
  • Can be used with the H8 Headgear easy faceshield replacement system
  • The shield goes inside the attachment for extra support
The WP98 Series clear acetate faceshield protects against high speed particles with low energy impact (0,86 g ball at 45 m/s), markingMore Product Information