Powered air respirator filter cover for Versaflo™ TR-6110E and TR-6130E, Versaflo™
Toimittaja: 3M
TR-6100FC
MMMATR-6100FCEA 28.9 EUR
MMMATR-6100FC
Powered air respirator filter cover for Versaflo™ TR-6110E and TR-6130E, Versaflo™
Hengityssuojaimet Hengityssuojajärjestelmät
The Versaflo™ Filter Cover TR-6100FC is a filter cover to help protect Versaflo™ TR-6110E and TR-6130E filters from physical damage.
  • Protect Versaflo™ TR-6110E and TR-6130E filters from physical damage
  • Holds a Versaflo™ Spark Arrestor, TR-662, or Versaflo™ Prefilter TR-6600 in place for further protection
This filter cover safeguards filters and enables users to wear Versaflo™ Powered Air Turbo through decontamination showers. It can hold a Versaflo™ Spark Arrestor TR-662, or Versaflo™ Prefilter TR-6600 in place for additional protection from largerMore Product Information