Safety-Walk™ 600 Slip-Resistant General-Purpose Tapes
Toimittaja: 3M
SW1SC50 SW1GB50 SW1TR50 SW1TR25 SW1GB25 SW1GB100 SW1TR100 SW1SC610 W1SC914N SW1SC150 SW1SC100 SW1SC19 SW1ST152 SW1SC25 SW1ST19N SW1SG25 SW1SG50 SW1SG150 SW1SG100 SW1SG305 SW1TR150N SW1TR305 SW1TR19 SW1GB19N SW1GB305 SW4ST153 SW1SC510N SW1SG610
MMMASW1SC50EA 237 EUR
MMMASW1SC50 MMMASW1GB50 MMMASW1TR50 MMMASW1TR25 MMMASW1GB25 MMMASW1GB100 MMMASW1TR100 MMMASW1SC610 MMMAW1SC914N MMMASW1SC150 MMMASW1SC100 MMMASW1SC19 MMMASW1ST152 MMMASW1SC25 MMMASW1ST19N MMMASW1SG25 MMMASW1SG50 MMMASW1SG150 MMMASW1SG100 MMMASW1SG305 MMMASW1TR150N MMMASW1TR305 MMMASW1TR19 MMMASW1GB19N MMMASW1GB305 MMMASW4ST153 MMMASW1SC510N MMMASW1SG610
Safety-Walk™ 600 Slip-Resistant General-Purpose Tapes
Teipit
3M™ Safety-Walk™ slip resistant general purpose 600 series tape is a mineral-coated, abrasive material adhered by a durable resin to a pressure-sensitive adhesive backed plastic film. It provides an effective slip-resistant surface in light to heavy shoe-traffic areas.
  • Anti-slip tape for light to heavy shoe traffic areas
  • High quality abrasive particles ensure long-lasting friction in any application
  • Highly durable surface withstands the rigors of equipment traffic
  • Low-profile design helps reduce trip hazard
  • High performance pressure-sensitive adhesive for