Safety glasses, SecureFit™ 500 series
Toimittaja: 3M
SF501AF-BLK SF501ASP-BLK SF501SGAF-BLK SF501SGAFBLKFM SF501SGAF-GRN SF501SGAF-RED SF502SGAF-BLK SF505SGAF-DGR SF507SGAF-BLK SF528SGAF-DGR
MMMASF501AF-BLKEA 14.5 EUR
MMMASF501AF-BLK MMMASF501ASP-BLK MMMASF501SGAF-BLK MMMASF501SGAFBLKFM MMMASF501SGAF-GRN MMMASF501SGAF-RED MMMASF502SGAF-BLK MMMASF505SGAF-DGR MMMASF507SGAF-BLK MMMASF528SGAF-DGR
Safety glasses, SecureFit™ 500 series
Suojalasit
The SecureFit™ 500 series safety glasses feature a stylised lens shape with temples that automatically self-adjust to help provide a secure and comfortable fit.
  • Sleek and compatible safety glasses with adjustable ratchet temples to adjust the lens for facial features and eyeglass position
  • Innovative SecureFit™ pressure diffusion temple technology provides comfortable pressure equalisation
  • Scotchgard™ anti-fog coating provides superior anti-fog and anti-scratch properties (available on selected models)
  • Protection against UV up to 400 nm
  • Wrap-around design