Nailsaver scouring sponges
Toimittaja: 3M
SCH105 SCH106 SCH363 SCH108 SCH107 SCH2276
MMMASCH105EA 32 EUR
MMMASCH105 MMMASCH106 MMMASCH363 MMMASCH108 MMMASCH107 MMMASCH2276
Nailsaver scouring sponges
Pyyhkeet, liinat ja sienet Sienet
3M™ Nailsaver scouring sponges are cleaning sponges made of a textured synthetic fibre white web containing very soft abrasive particles, laminated with an adhesive onto a grooved synthetic colour-coded foam holder.
  • Easy positioning four colours of foams) to comply with HACCP requirements
  • Good cleaning performance for light-duty cleaning tasks
  • Soft and low scratching power on most surfaces
  • Easy to handle and finger protection thanks to the shape of the foam
  • Very long lasting
For easy cleaning with low scratching power on most surfaces, use 3M™More Product Information