CCS safety glasses, Solus™
Toimittaja: 3M
SCCS01SGAF-GRN SCCS01SGAFGRNF
MMMASCCS01SGAF-GRNEA 13.7 EUR
MMMASCCS01SGAF-GRN MMMASCCS01SGAFGRNF
CCS safety glasses, Solus™
Suojalasit
The Solus™ CCS safety glasses feature a rimless design with lime green temples and a corded control system (CCS) for attaching corded earplugs. The polycarbonate lens features a Scotchgard™ anti-fog and anti-scratch coating on both sides for increased durability and improved vision.
  • Modern safety glasses range keeps the eyewear and corded earplugs attached and ready to use
  • Scotchgard™ anti-fog coating provides superior anti-fog and anti-scratch properties (available on selected models)
  • Suitable for use with all non-textile 3M™ corded earplugs
  • Available