Full Facepiece Reusable Respirators, FF-600F
Toimittaja: 3M
FF-601F FF-602F FF-603F
MMMAFF-601FEA 397 EUR
MMMAFF-601F MMMAFF-602F MMMAFF-603F
Full Facepiece Reusable Respirators, FF-600F
Hengityssuojaimet Uudelleen käytettävät maskit Kokomaskit
3M™ reusable full face mask, FF-600 series features a panoramic visor that allows for a virtually unrestricted field of vision. Compatible with the 3M™ filters, DT series, the FF-600 series is suitable for a wide range of wearers and applications.
  • Panoramic visor to allow for a virtually unrestricted field of vision
  • Visor is designed to minimise visual distortion
  • Soft, highly flexible and conforming liquid silicone rubber faceseal to enhance user comfort
  • Speech diaphragm
  • Front port filter
  • Three sizes are available
  • Facepiece weight: approx. 550 g