High shine double sided floor pads, Scotch-Brite™
Toimittaja: 3M
CLSH430
MMMACLSH430EA 1260 EUR
MMMACLSH430
High shine double sided floor pads, Scotch-Brite™
Pyyhkeet, liinat ja sienet Puhdistuspakkaukset ja -tyynyt
Scotch-Brite™ High Shine Double Sided Floor Pads cut costs without compromising shine. Not only does it clean and polish marble, terrazzo, plastic, and concrete floors to a brilliant gloss finish, it goes on working for three times longer than comparable floor pads, resulting in significant total cost savings.
  • High Shine Floor Pads for floors with medium shine in good initial condition; cleans and polishes floors to a brilliant gloss finish
  • Precision engineered to last three times longer than existing single-sided high performance floor pads
  • Double-sided