Double Coated Urethane Foam Tapes, 4026
Toimittaja: 3M
40261933 40261233 40262533 40263833
MMMA40261933EA 93.3 EUR
MMMA40261933 MMMA40261233 MMMA40262533 MMMA40263833
Double Coated Urethane Foam Tapes, 4026
Teipit
3M™ 4026 double coated urethane foam tape is a double-sided, urethane foam tape that provides a strong and reliable bond on a variety of surfaces with a high shear strength. It's designed to fill gaps and distribute stress uniformly over the bonded area.
  • Durable foam is ideal for interior mounting, bonding, and attaching applications
  • Open-cell, flexible urethane foam carrier dampens and prevents vibration
  • Provides good insulating qualities
  • High shear adhesive on metals and plastics with high temperature resistance
  • Designed for indoor use
  • Suitable for soft