Flexible fit probed test earplugs, E-A-R™
Toimittaja: 3M
393-2026-50
MMMA393-2026-50EA 0 EUR
MMMA393-2026-50
Flexible fit probed test earplugs, E-A-R™
Korvatulpat Kertakäyttöiset korvatulpat
E-A-R™ Flexible Fit Earplugs equipped with a probe tube for fit testing using the 3M™ E-A-Rfit™ Dual-Ear Validation System. Cannot be used for hearing protection.
  • Test earplugs are modified with a probe tube for use during earplug fit testing
  • 50 pairs per case
  • For occupational use only
These test Plugs are a specially probed version of the standard earplug. This product works with the 3M™ E-A-Rfit™ Dual-Ear Validation System to allow fit tests of Flexible Fit earplugs.