High performance cleaning wipes, 2011 series, Scotch-Brite™
Toimittaja: 3M
2011HLGN 2011HLRT 2011AUTO
MMMA2011HLGNEA 0 EUR
MMMA2011HLGN MMMA2011HLRT MMMA2011AUTO
High performance cleaning wipes, 2011 series, Scotch-Brite™
Pyyhkeet, liinat ja sienet Uudelleenkäytettävät puhdistusliinat Liinat yleiskäyttöön
The 2011 series Scotch-Brite™ high performance cleaning wipes can be used dry or damp, leaving a lint-free, streak-free finish.
  • Wipes can be used dry or damp, leaving a lint-free, streak-free finish
  • Eliminates the need for cleaning agents in many cases
  • Machine washable up to 75 times at 95 °C
Clean any surface with Scotch-Brite™ high performance cleaning wipes, 2011 series. These long lasting, high performance cleaning wipes are ideal for removing dust, dirt, grease, and fingerprints from stainless steel, chrome, plastics, glass, paint lacquers, andMore Product Information