Aluminium foil tapes, 1436
Toimittaja: 3M
1436-100MM 1436-50MM 1436-75MM
MMMA1436-100MMEA 447 EUR
MMMA1436-100MM MMMA1436-50MM MMMA1436-75MM
Aluminium foil tapes, 1436
Teipit
3M™ aluminum foil tape 1436 is a high-quality tape for internal and external ducting work. It offers an aggressive adhesive for ease of application.
  • Foil backing provides an excellent reflective surface for both heat and light
  • Low water vapour transmission rate offers excellent sealing and patching performance
  • Performance range: –25 to 70 °C (short term 100 °C)
  • Very high initial tack to most surfaces
  • Easy release liner
  • Tensile strength: 45 [Newton/25 mm]
  • Dead soft aluminium back material
It has an aggressive rubber adhesive and is supplied with easyMore Product Information