Western blot assay kits, ScanLater™
Toimittaja: MOLECULAR DEVICES
R8200 R8201 R8202 R8203 R8204 R8205 R8207 R8206 R8208 R8209 R8212 R8228 R8224 R8225 R8226 R8227
MLDVR8200EA 0 EUR
MLDVR8200 MLDVR8201 MLDVR8202 MLDVR8203 MLDVR8204 MLDVR8205 MLDVR8207 MLDVR8206 MLDVR8208 MLDVR8209 MLDVR8212 MLDVR8228 MLDVR8224 MLDVR8225 MLDVR8226 MLDVR8227
Western blot assay kits, ScanLater™
Elektroforeesireagenssit Western Blotting Reagents
The ScanLater™ Western Blot Assay Kits are a time-resolved fluorescence (TRF)-based assay, optimal for quantitating as little as femtogram protein samples.
  • Time-resolved fluorescence detection
  • Europium-labeled goat, anti-rabbit, anti-mouse or streptavidin labeled antibodies
  • Supports nitrocellulose and PVDF membranes
  • 340/80 nm excitation with 616/10 nm emission range
  • Optimised for use with ScanLater™ Western Blot Detection Cartridge for the SpectraMax® i3 and Paradigm® Multi-Mode Platforms
Using this Western blot method, researchers can eliminate time-dependent substrate addition steps and sustain blot signal stability for at least one month. Researchers can also enhance assay sensitivity using TRF method to reduce background noise. This kit allows researchers to maintain femtogram to picogram protein sensitivity, similar to traditional western blot detectors.
Toimitustiedot: Evaluation kit includes 20 µg antibody, 40 ml blocking buffer, 100 ml wash buffer (for 10 mini gels with recommended protocol). Bulk kit includes 60 µg antibody, 120 ml blocking buffer, 300 ml wash buffer (for 30 mini gels with recommended protocol).