CloneMatrix
Toimittaja: MOLECULAR DEVICES
K8510 K8530
MLDVK8510EA 0 EUR
MLDVK8510 MLDVK8530
CloneMatrix
Soluviljelyyn liuokset
CloneMatrix is a 2,5X concentrate that serves as a matrix to produce semi-solid media for hybridoma, myeloma, CHO, HEK, and mESC.