Filter holders, stainless steel
Toimittaja: Merck
YY3009000 YY3014236
MILFYY3009000EA 2050 EUR
MILFYY3009000 MILFYY3014236
Filter holders, stainless steel
Suodatintelineet
Standing filter holders with anodised aluminium legs for contamination analysis.
  • With PTFE O-rings
  • ¹/₈" NPTM vent/relief valve attachment