Chemiluminescent detection reagents, Immobilon® Western HRP substrates
Toimittaja: Merck
WBKLS0100 WBKLS0500 WBKLS0050
MILFWBKLS0100EA 171 EUR
MILFWBKLS0100 MILFWBKLS0500 MILFWBKLS0050
Chemiluminescent detection reagents, Immobilon® Western HRP substrates
Elektroforeesireagenssit Western Blotting Reagents
Immobilon® Western HRP substrates are compatible with both PVDF and nitrocellulose membranes, as well as all commonly used buffers and blocking reagents.