Centrifugal filters, Amicon® Ultra-0,5 ml
Toimittaja: Merck
UFC501024 UFC501096 UFC510096 UFC503008 UFC500308 UFC501008 UFC505008 UFC503096 UFC500324 UFC500396 UFC503024 UFC510024 UFC505024 UFC510008 UFC5003BK UFC5010BK UFC5030BK UFC505096 UFC5050BK UFC5100BK
MILFUFC501024EA 161 EUR
MILFUFC501024 MILFUFC501096 MILFUFC510096 MILFUFC503008 MILFUFC500308 MILFUFC501008 MILFUFC505008 MILFUFC503096 MILFUFC500324 MILFUFC500396 MILFUFC503024 MILFUFC510024 MILFUFC505024 MILFUFC510008 MILFUFC5003BK MILFUFC5010BK MILFUFC5030BK MILFUFC505096 MILFUFC5050BK MILFUFC5100BK
Centrifugal filters, Amicon® Ultra-0,5 ml
Suodattimet Sentrifugisuodattimet
Ultra centrifugal filters provide fast sample processing and promote high sample recoveries, even in dilute samples, through ultrafiltration.
  • Heat-sealed membrane eliminates adhesives and downstream extractables
  • Enables maximum protein recovery without introducing pipetting errors
  • Compatible with pH 1 to 9
  • Low binding membrane and PP housing for > 90% sample recovery
The unique features of the Amicon® Ultra centrifugal filters gives the fastest, most efficient concentration for sensitive downstream applications.