Membrane filters, hydrophobic edge
Toimittaja: Merck
GSEP047S0 HAEG047AW HAEG047S0 SMEP0470W
MILXGSEP047S0EA 0 EUR
MILXGSEP047S0 MILXHAEG047AW MILXHAEG047S0 MILFSMEP0470W
Membrane filters, hydrophobic edge
Suodattimet Membraanisuodattimet
Hydrophobic edge membranes are standard MF-type filters with edges rendered hydrophobic to 3 or 6 mm widths. In antibiotic and bacteriostatic drug sterility testing by membrane filter method, the 6 mm wide non-wetting edge prevents drug intrusion under holder sealing rim.
  • Available in sterile or autoclave as ready packs
Applications include microbiological and sterility testing of antibiotics and bacteriostatic drugs and filling machine operations.
Filter surface: plain
Filter color: white