Millex™ HV Syringe Filter
Toimittaja: Merck
SLHV013NK
MILFSLHV013NKEA 1900 EUR
MILFSLHV013NK
Millex™ HV Syringe Filter
Suodattimet Ruiskusuodattimet
Millex-HV Syringe Filter with PVDF Membrane bind far less protein than nylon, nitrocellulose membranes, or PTFE membranes. High flow rates and throughput, low levels of extractables and broad chemical compatibility.
  • Polypropylene housing
  • Low protein binding to minimise interaction with sample and maximise recovery
  • Used in preparation of protein-containing solutions prior to chromatography or other instrument analyses
  • Filter Code: HV
Chemistry: Hydrophilic Polyvinylidene Fluoride (PVDF).