Millex® MCE MF-Millipore™ Membrane Syringe Filters, Sterile
Toimittaja: Merck
SLAAR33SB SLAAR33SS SLGSR33SB SLGSR33SS SLHAR33SS
MILFSLAAR33SBEA 695 EUR
MILFSLAAR33SB MILFSLAAR33SS MILFSLGSR33SB MILFSLGSR33SS MILFSLHAR33SS
Millex® MCE MF-Millipore™ Membrane Syringe Filters, Sterile
Suodattimet Ruiskusuodattimet
Sterile Millex filter units are available in a larger 33 mm housing with MCE membranes.
The larger filter surface area increases flow rate and throughput. It also makes it easier to filter solutions because it reduces the pressure required to empty the syringe. Millex filters have a maximum housing pressure of 10,5 bar, which means you can filter solutions faster than before.
The colour-coded band on the Millex housing clearly indicates which membrane is inside. Millex filters are manufactured in a controlled environment using an automated process. Human hands never touch the filterMore Product Information