Multiscreen® 96-Well Plates with Polycarbonate Membrane
Toimittaja: Merck
MAMIC3S10 MAMIC8S10 MAMIC5S10
MILFMAMIC3S10EA 1140 EUR
MILFMAMIC3S10 MILFMAMIC8S10 MILFMAMIC5S10
Multiscreen® 96-Well Plates with Polycarbonate Membrane
Kuoppalevyt Suodatinlevyt
HTS filter plates (Plate catalogue numbers that start with MS): MultiScreenHTS plates are specifically developed for high-throughput use with automated work stations. Rigid sidewalls improve handling and ample surfaces are provided for bar code labels. Wells are individually sealed to prevent incubation crosstalk and the underdrain is removable for access to the filters. Filter plates are available in 96- and 384-well platforms with a broad selection of membranes and plastics. Millipore's new MultiScreen HTS+ Hi flow filter plate design lessens non-specific binding and reduces variability inMore Product Information