Montage® Gel Extraction Kit
Toimittaja: Merck
LSKGEL050 LSKMTAE50
MILFLSKGEL050EA 0 EUR
MILFLSKGEL050 MILFLSKMTAE50
Montage® Gel Extraction Kit
Nukleiinihapporeagenssit Nucleic Acid Purification Kits and Reagents
The device utilises gel compression to extract DNA from the agarose. Centrifugal force collapses the gel structure, drives the agarose through a small orifice in the gel nebulizer and the resultant gel slurry is sprayed into the sample filter cup.
  • Minimal hands-on time
  • 10-minute spin
  • Fully-functional DNA
Recover 100 to 10000 bp DNA from agarose gel slices in a single 10-minute spin. The kit consists of a pre-assembled filter device with an agarose gel nebulizer, a microcentrifuge vial, and modified TAE gel extraction buffer (dry).
Prepared DNA requires no further purification for most applications, including cloning and radioisotopic or fluorescent DNA sequencing. Since agarose gel electrophoresis has high resolving power, the small and large non-specific amplification products that frequently interfere with cloning and sequencing after PCR are completely removed from the product.
Application: Genomic analysis
Toimitustiedot: Includes 50 Ultrafree-DA devices and 500 ml of 50X concentrated modified TAE buffer.