Visualizer Western Blot Detection Kit

Toimittaja: Merck

64-201 64-202BP 64-202 64-201BP
MILF64-201EA 534 EUR
MILF64-201 MILF64-202BP MILF64-202 MILF64-201BP
Visualizer Western Blot Detection Kit
Elektroforeesireagenssit Western Blotting Reagents

The Visualizer Western Blot Detection kit is designed for the sensitive detection of proteins in chemiluminescent Western blots.


The reagents in this kit have been matched to optimise the range and sensitivity of detection using various antibodies in Western blot assays.


Components: Goat anti-mouse IgG, goat anti-rabbit IgG, HRP conjugate Visualizer detection reagent A Visualizer detection reagent B


Storage: Stable for 18 months at 4 °C
Application: For use on immunoblots

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR