Autosampler vials, inserts and kits, RSA™
Toimittaja: MICROSOLV
9509S-0CV-RS 9509S-1WCP-RS 9509C-1EWCP-RS 9509C-1WCP-RS 9509C-1XWCP-RS 9509C-3EWCP-RS 9509C-3WAP-RS 9509C-3WCP-RS 9509C-3XWAP-RS 9509C-3XWCP-RS 9509C-WAV-RS 9509C-WCV-RS 9509S-1CP-RS 9509S-1EWAP-RS 9509S-1EWCP-RS 9509S-1WAP-RS 9509S-1WCP-RSA 9509S-1XWAP-RS 9509S-1XWCP-RS 9509S-3CP-RS 9509S-3EWAP-RS 9509S-3EWCP-RS 9509S-3WAP-RS 9509S-3WCPEARS 9509S-3WCP-RS 9509S-3XWAP-RS 9509S-3XWCP-RS 9509S-9WAP-RS 9509S-9WCP-RS 9509S-WAV-RS 9509S-WCV-RS 9512C-0CV-RS 9512C-0CV-T-RS 9512C-1CP-RS 9512C-1MP-RS 9512C-3CP-RS 9512C-3MP-RS 9512S-0CV-RS 9512S-1AP-RS 9512S-1CP-RS 9512S-1MP-RS 9512S-3AP-RS 9512S-3CP-RS 9512S-3MP-RS 9512S-9AP-RS 9512S-9CP-RS 9532C-0CV-RS 9532C-1CP-RS 9532C-3CP-RS 9532S-0CV-RS 9532S-1CP-RS 9532S-1CPWT-RS 9532S-3CP-RS 9532S-3CPWT-RS 9532S-WCV-WTRS 9512S-0CV-T-RS
MICS9509S-0CV-RSEA 34.4 EUR
MICS9509S-0CV-RS MICS9509S-1WCP-RS MICS9509C-1EWCP-RS MICS9509C-1WCP-RS MICS9509C-1XWCP-RS MICS9509C-3EWCP-RS MICS9509C-3WAP-RS MICS9509C-3WCP-RS MICS9509C-3XWAP-RS MICS9509C-3XWCP-RS MICS9509C-WAV-RS MICS9509C-WCV-RS MICS9509S-1CP-RS MICS9509S-1EWAP-RS MICS9509S-1EWCP-RS MICS9509S-1WAP-RS MICS9509S-1WCP-RSA MICS9509S-1XWAP-RS MICS9509S-1XWCP-RS MICS9509S-3CP-RS MICS9509S-3EWAP-RS MICS9509S-3EWCP-RS MICS9509S-3WAP-RS MICS9509S-3WCPEARS MICS9509S-3WCP-RS MICS9509S-3XWAP-RS MICS9509S-3XWCP-RS MICS9509S-9WAP-RS MICS9509S-9WCP-RS MICS9509S-WAV-RS MICS9509S-WCV-RS MICS9512C-0CV-RS MICS9512C-0CV-T-RS MICS9512C-1CP-RS MICS9512C-1MP-RS MICS9512C-3CP-RS MICS9512C-3MP-RS MICS9512S-0CV-RS MICS9512S-1AP-RS MICS9512S-1CP-RS MICS9512S-1MP-RS MICS9512S-3AP-RS MICS9512S-3CP-RS MICS9512S-3MP-RS MICS9512S-9AP-RS MICS9512S-9CP-RS MICS9532C-0CV-RS MICS9532C-1CP-RS MICS9532C-3CP-RS MICS9532S-0CV-RS MICS9532S-1CP-RS MICS9532S-1CPWT-RS MICS9532S-3CP-RS MICS9532S-3CPWT-RS MICS9532S-WCV-WTRS MICS9512S-0CV-T-RS
Autosampler vials, inserts and kits, RSA™
Näytevialit ja pienet näyteastiat
RSA™ (Reduced Surface Activity) high quality and extremely consistent autosampler vials and caps due to reduced surface activity glass manufacturing and control processes. Excellent for low abundance or low dosage form analytes or samples that are pH sensitive, or basic in nature.
  • Low adsorption glass, useful for low abundance basic compounds
  • No pH change from the vial when using a high amount of water in samples
  • Virtually no surface metals prevents adducts in LCMS
  • Completely manufactured robotically, not touched by human hands
  • Reliability and conistency