HPLC columns, Cogent Diamond Hydride™
Toimittaja: MICROSOLV
70000-05P-2 70000-15P 70200-05P-2 70000-7.5P 70000-P30-100 70200-10P-2 70000-HG9 70000-05P 70000-05P-1 70000-10P 70000-10P-1 70000-10P-2 70000-15P-1 70000-15P-2 70000-25P 70000-HG5 70000-HG6 70000-P21-100 70000-P21-150 70000-P21-250 70000-P21-50 70000-P30-250 70000-P30-50 70000-SP100 70000-SP150 70000-SP250 70000-SP50 70200-10P-1 70200-15P-1 70200-15P-2 70200-HG3
MICS70000-05P-2EA 933 EUR
MICS70000-05P-2 MICS70000-15P MICS70200-05P-2 MICS70000-7.5P MICS70000-P30-100 MICS70200-10P-2 MICS70000-HG9 MICS70000-05P MICS70000-05P-1 MICS70000-10P MICS70000-10P-1 MICS70000-10P-2 MICS70000-15P-1 MICS70000-15P-2 MICS70000-25P MICS70000-HG5 MICS70000-HG6 MICS70000-P21-100 MICS70000-P21-150 MICS70000-P21-250 MICS70000-P21-50 MICS70000-P30-250 MICS70000-P30-50 MICS70000-SP100 MICS70000-SP150 MICS70000-SP250 MICS70000-SP50 MICS70200-10P-1 MICS70200-15P-1 MICS70200-15P-2 MICS70200-HG3
HPLC columns, Cogent Diamond Hydride™
Kromatografiakolonnit
These useful HPLC columns have a silica hydride surface similar to other TYPE-C™ phases but the uniqueness of Diamond Hydride™ is that they have a very small amount of carbon on the surface that adjusts the hydrophobicity to a small but important level. This is an excellent choice for small, polar compounds by LCMS or HPLC.
  • No water shell
  • Fast equilibration in HILIC mode
  • Excellent for polar compounds and some non-polar compounds
Popular choice of scientists working in metabolomics or bioanalytical with LCMS for compounds such as amino acids,More Product Information