HPLC columns, Cogent Phenyl Hydride™
Toimittaja: MICROSOLV
69020-25P 69020-05P 69020-05P-1 69020-05P-2 69020-10P 69020-10P-1 69020-10P-2 69020-15P 69020-15P-1 69020-15P-2 69020-HG5 69020-P21-100 69020-P21-150 69020-P21-250 69020-P21-50 69020-P30-100 69020-P30-150 69020-P30-250 69020-P30-50 69020-SP100 69020-SP150 69020-SP250 69020-SP50 69220-05P-1 69220-05P-2 69220-10P-1 69220-10P-2 69220-15P-2
MICS69020-25PEA 1340 EUR
MICS69020-25P MICS69020-05P MICS69020-05P-1 MICS69020-05P-2 MICS69020-10P MICS69020-10P-1 MICS69020-10P-2 MICS69020-15P MICS69020-15P-1 MICS69020-15P-2 MICS69020-HG5 MICS69020-P21-100 MICS69020-P21-150 MICS69020-P21-250 MICS69020-P21-50 MICS69020-P30-100 MICS69020-P30-150 MICS69020-P30-250 MICS69020-P30-50 MICS69020-SP100 MICS69020-SP150 MICS69020-SP250 MICS69020-SP50 MICS69220-05P-1 MICS69220-05P-2 MICS69220-10P-1 MICS69220-10P-2 MICS69220-15P-2
HPLC columns, Cogent Phenyl Hydride™
Kromatografiakolonnit
Cogent HPLC columns have a silicon hydride surface similar to other TYPE-C™ phases but the uniqueness of Phenyl Hydride™ is that they have unique selectivity for compounds with aromatic rings with all the TYPE-C silica benefits.
  • Excellent for Aromatic Compounds 
  • Use in RP or ANP
  • TYPE-C™ Silica
Direct silicon carbon bonds and the silica hydride surface give these columns extremely long life times compared to other phenyl columns based on ordinary silica.
Used also with LCMS because there is no endcapping that will bleed into your MS as withMore Product Information