Inspection mirror
Toimittaja: MICRONOVA
QDIST-66P
MICRQDIST-66PEA 528 EUR
MICRQDIST-66P
Inspection mirror
Peilit
Inspection Mirror has a swivel connection allowing it to be attached to stainless steel handle and allows the operator to run the unit behind equipment and fixtures to ensure all surfaces have been properly cleaned.
  • Highly polished stainless steel
  • A 360° swivel allows optimum flexibility
  • Ideal for hard to reach areas
Tilaustiedot: The optional flashlight attachment is ideal for use in tanks and other low light applications. Rod is not included, but fits to any SSU series handle.